Skip to Content
MilliporeSigma

MAK115

Hemoglobin Assay Kit

sufficient for 250 colorimetric tests

Synonym(s):

Hemoglobin Quantitation Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.84
UNSPSC Code:
12161503
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


usage

sufficient for 250 colorimetric tests

species reactivity

rat, human, mouse

packaging

pkg of 1 kit

storage condition

dry at room temperature

technique(s)

activity assay: suitable

input

plasma
urine
blood
serum

accession no.

P69905, NM_000558.5

detection method

colorimetric

relevant disease(s)

hematological disorder

storage temp.

2-8°C

Gene Information

human ... HBA1(3039), HBA2(3040), HBB(3043)
mouse ... HBB(15129)
rat ... HBB(24440)

General description

Hemoglobin (Hb) is an iron-containing metalloprotein that serves as the primary means of oxygen transport in vertebrates. Hemoglobin is primarily found in red blood cells where it makes up to 97% of the cell′s dry content. Hemoglobin can also be found in other tissues where it serves as an antioxidant. Alterations in blood hemoglobin levels occurs in many diseases such as anemia and polycytheima.

Application

Hemoglobin Assay Kit has been used to measure the concentration of hemoglobin.
Suitable for the detection of hemoglobinin a variety of samples such as blood, serum, plasma, and urine. This kit can be used to study the effects of various compounds on hemoglobin metabolism and levels.

Biochem/physiol Actions

This assay is based on the improved Triton[NP1]/NaOH method in which hemoglobin is converted to a colorimetric product measured at 400 nm. This assay has a linear detection range between 0.9-200 mg/dL in the 96 well plate assay.

Features and Benefits

Compatible with high-throughput handling systems. Can be adapted for use with cuvettes.

Legal Information

Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow


pictograms

CorrosionEnvironment

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - ED ENV 1 - Eye Irrit. 2 - Met. Corr. 1

Storage Class

12 - Non Combustible Liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library