Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MAK059

Sigma-Aldrich

Formate Assay Kit

sufficient for 100 colorimetric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

usage

sufficient for 100 colorimetric tests

application(s)

cosmetics
environmental
food and beverages

detection method

colorimetric

storage temp.

−20°C

General description

Formate, the salt form of formic acid, is the simplest carboxylate anion. Formate is produced in the mitochondria as a result of the oxidation of one-carbon donors such as serine. Mitochondrially-derived formate is utilized for cytoplasmic one-carbon metabolism in the de novo synthesis of purines, thymdiylate, and the remethylation of homocysteine to methionine. Under normal physiologic conditions, formate is present at low levels in serum and blood but levels may be increased following acute methanol poisoning or environmental exposure to formaldehyde. Serum and urine formate levels were increased in vitamin B12-deficient rats and may be indicative of hyperhomocysteinemia due to defects in remethylation.

Application

Formate Assay kit has been used to determine the concentration of formate in samples.

Suitability

Suitable for serum, urine, media, cells, and tissues.

Principle

The Formate Assay Kit provides a simple, sensitive, and rapid means of quantifying formate in a variety of samples including serum, urine, media, cells, and tissues. Formate concentrations are determined by an enzymatic assay, which results in a colorimetric (450 nm) product, proportional to the formate present.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Providing reducing power by microalgal photosynthesis: a novel perspective towards sustainable biocatalytic production of bulk chemicals exemplified for aliphatic amines
Lowe J, et al.
Scientific Reports, 8, 10436-10436 (2018)
Simon G Lamarre et al.
American journal of physiology. Endocrinology and metabolism, 302(1), E61-E67 (2011-09-22)
Formate can differentiate between hyperhomocysteinemia due to impaired remethylation and impaired transsulfuration. Am J Physiol Endocrinol Metab 301: E000-E000, 2011. First published September 20, 2011; 10.1152/ajpendo.00345.2011.-We carried out a (1)H-NMR metabolomic analysis of sera from vitamin B(12)-deficient rats. In addition
Benjamin J Koestler et al.
mBio, 9(5) (2018-09-27)
The intracellular human pathogen Shigella flexneri invades the colon epithelium, replicates to high cell density within the host cell, and then spreads to adjacent epithelial cells. When S. flexneri gains access to the host cytosol, the bacteria metabolize host cytosolic
Henning Kirst et al.
Proceedings of the National Academy of Sciences of the United States of America, 119(8) (2022-02-24)
Formate has great potential to function as a feedstock for biorefineries because it can be sustainably produced by a variety of processes that don't compete with agricultural production. However, naturally formatotrophic organisms are unsuitable for large-scale cultivation, difficult to engineer
Cornelia U Welte et al.
Environmental microbiology, 23(7), 4017-4033 (2021-04-30)
Methoxylated aromatic compounds (MACs) are important components of lignin found in significant amounts in the subsurface. Recently, the methanogenic archaeon Methermicoccus shengliensis was shown to be able to use a variety of MACs during methoxydotrophic growth. After a molecular survey

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service