Skip to Content
MilliporeSigma

M9187

Greiner CELLSTAR® multiwell culture plates

12 wells (suspension culture with lid)

Synonym(s):

culture dishes, culture plates, multi dishes, Greiner CELLSTAR® multiwell culture plates, Greiner 665102

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.22
UNSPSC Code:
41122107
Material:
flat clear wells, polystyrene
Size:
12 wells
Sterility:
sterile
Binding type:
non-treated surface
Feature:
lid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

flat clear wells, polystyrene

sterility

sterile

feature

lid

packaging

pack of 1 ea (single packed)

manufacturer/tradename

Greiner 665102

size

12 wells

well volume

6.5 mL

well working volume

2- 4 mL

binding type

non-treated surface

Features and Benefits

  • CELLSTAR® 12 Well Cell Culture Multiwell Plates with a hydrophobic surface is ideal for suspension culture cells and hybridoma cells.
  • Features a lid that facilitates gas exchange with the lowest possible evaporation and significantly reduces the risk of cross-contamination.
  • Free of detectable DNase, RNase, human DNA, non-pyrogenic.
  • Suitable for all common instruments and compatible with automated systems.
  • Alphanumeric well coding for easy well/sample identification.

Legal Information

CELLSTAR is a registered trademark of Greiner Bio-One GmbH


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library