Skip to Content
MilliporeSigma

M8449

MAN-5 Glycan

≥90% (HPLC)

Synonym(s):

(Man)5(GlcNAc)2, Mannopentaose-di-(N-acetyl-D-glucosamine), Oligomannose-5 glycan

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C46H78N2O36
CAS Number:
Molecular Weight:
1235.10
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352201
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥90% (HPLC)

form

solid

storage temp.

−20°C

SMILES string

CC(=O)NC(C=O)C(O)C(OC1OC(CO)C(OC2OC(COC3OC(COC4OC(CO)C(O)C(O)C4O)C(O)C(OC5OC(CO)C(O)C(O)C5O)C3O)C(O)C(OC6OC(CO)C(O)C(O)C6O)C2O)C(O)C1NC(C)=O)C(O)CO

InChI

1S/C46H78N2O36/c1-11(55)47-13(3-49)22(58)37(14(57)4-50)81-41-21(48-12(2)56)28(64)38(18(8-54)78-41)82-46-36(72)40(84-45-34(70)31(67)25(61)17(7-53)77-45)27(63)20(80-46)10-74-43-35(71)39(83-44-33(69)30(66)24(60)16(6-52)76-44)26(62)19(79-43)9-73-42-32(68)29(65)23(59)15(5-51)75-42/h3,13-46,50-54,57-72H,4-10H2,1-2H3,(H,47,55)(H,48,56)

InChI key

KIAGZJJOMMNKBW-UHFFFAOYSA-N

Biochem/physiol Actions

Antibodies bearing high levels of N-linked mannose-5 glycan (Man-5) have been found to exhibit enhanced antibody-dependent cell-mediated cytotoxicity (ADCC) when compared to antibodies with fucosylated complex or hybrid glycans.
Antibodies carrying high levels of N-linked mannose-5 glycan (Man5) possesses increased antibody-dependent cell-mediated cytotoxicity (ADCC) than antibodies with fucosylated complex or hybrid glycans.

Other Notes

To gain a comprehensive understanding of our extensive range of Oligosaccharides for your research, we encourage you to visit our Carbohydrates Category page.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library