Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

M4436

Greiner high and medium binding 96 well plates

flat bottom clear wells

Synonym(s):

96 multiwell plates, 96 well microplates, 96 well microtiter plates, 96 well strip plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
NACRES:
NB.22
material:
clear polystyrene
flat bottom clear wells
size:
96 wells
well volume:
382 μL
well working volume:
25-340 μL
sterility:
non-sterile
binding type:
medium binding surface
feature:
lid: no
skirt (F-bottom)

material

clear polystyrene
flat bottom clear wells

sterility

non-sterile

feature

lid: no
skirt (F-bottom)

packaging

case of 40

manufacturer/tradename

Greiner 655001

size

96 wells

well volume

382 μL

well working volume

25-340 μL

binding type

medium binding surface

Looking for similar products? Visit Product Comparison Guide

General description

MICROLON® 200 96W Microplate, well working volume 25-340 μL; total well volume 382 μL.
Polystyrene Multiwell plates from Greiner for immunoassay and high throughput screening applications. High binding plates are recommended for ELISAs. High binding denotes a binding capacity of 600 ng/cm2 and scatter of less than 10%. Optimal protein binding is variable and comparative tests between high and medium binding plates is suggested.

Other Notes

96 well flat (F) bottom medium binding microplate, without lid, ; alphanumeric well coding, length: 127.76 mm; width: 85.48 mm; curvature approx. 200 um.

Suitability

UsefμL in immunological assays. MICROLON 200 is a hydrophobic medium binding surface that will bind up to 200 ng/cm2 of non-polar proteins and peptides. Free of detectable DNase/RNase, human DNA and pyrogens.

Legal Information

MICROLON is a registered trademark of Greiner Bio-One GmbH

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Assay protocol for the colorimetric detection of Branched Chain Amino Acids (BCAA) in biological samples using the alanine assay kit.

Protocols

Assay protocol for the colorimetric detection of Hydroxyproline in biological samples using the Hydroxypoline assay kit.

Assay protocol for the colorimetric detection of Hydroxyproline in biological samples using the Hydroxypoline assay kit.

Assay protocol for the colorimetric detection of Hydroxyproline in biological samples using the Hydroxypoline assay kit.

Assay protocol for the colorimetric detection of Hydroxyproline in biological samples using the Hydroxypoline assay kit.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service