Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M3449

Sigma-Aldrich

Mephedrone hydrochloride

Synonym(s):

4-Methylmethcathinone hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15NO·HCl
CAS Number:
Molecular Weight:
213.70
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

drug control

USDEA Schedule I; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IIA (Portugal)

color

white to off-white

solubility

DMSO: >20 mg/mL

storage temp.

room temp

SMILES string

Cl.CNC(C)C(=O)c1ccc(C)cc1

InChI

1S/C11H15NO.ClH/c1-8-4-6-10(7-5-8)11(13)9(2)12-3;/h4-7,9,12H,1-3H3;1H

InChI key

DLQZFTUKJGRPLZ-UHFFFAOYSA-N

Related Categories

Biochem/physiol Actions

Mephedrone is a synthetic stimulant, an analog of Cathinone and Methamphetamine.
Mephedrone is an inhibitor of monoamine transporter present in the brain and periphery. Mephedrone bears a chiral center and can be found in two enantiomers R-mephedrone and S-mephedrone. Its racemic form is often used as a drug. Mephedrone in rats is known to cause high blood pressure and heart rate, as well as stimulates locomotor hyperactivity. Human trail reveal that mephedrone has the ability to induce euphoria and pleasure.

Features and Benefits

This compound is featured on the Dopamine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neuropharmacology, 318-318 (2017)

Articles

We offer many products related to dopamine receptors for your research needs.

We offer many products related to dopamine receptors for your research needs.

We offer many products related to dopamine receptors for your research needs.

We offer many products related to dopamine receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service