Skip to Content
MilliporeSigma

M2398

Metaproterenol hemisulfate salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C11H17NO3 · 1/2H2SO4
CAS Number:
Molecular Weight:
260.30
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352107
EC Number:
227-539-6
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

originator

Boehringer Ingelheim

SMILES string

OS(O)(=O)=O.CC(C)NCC(O)c1cc(O)cc(O)c1.CC(C)NCC(O)c2cc(O)cc(O)c2

InChI

1S/2C11H17NO3.H2O4S/c2*1-7(2)12-6-11(15)8-3-9(13)5-10(14)4-8;1-5(2,3)4/h2*3-5,7,11-15H,6H2,1-2H3;(H2,1,2,3,4)

InChI key

MKFFGUZYVNDHIH-UHFFFAOYSA-N

Gene Information

human ... ADRB2(154)

Application

Metaproterenol hemisulfate salt has been used:
  • as a β2-adrenoceptor agonist to study its effects on adenylyl cyclase stimulation in red blood cell deformability
  • as an adrenergic agonist to study its effects on the stereoselective cell uptake by organic cation transporters in human embryonic kidney cell lines
  • as a bronchodilator to study its effects on the activation of G-protein coupled receptors (GPCR)

Biochem/physiol Actions

Metaproterenol is a β2-adrenoceptor agonist. It shows bronchodilator activity and shows therapeutic effects against bronchitis, asthma, and emphysema.

Features and Benefits

This compound was developed by Boehringer Ingelheim. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Inhalation

Storage Class

11 - Combustible Solids

wgk

WGK 2

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library