Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M168

Sigma-Aldrich

Methyllycaconitine citrate salt

from Delphinium brownii seeds, ≥96% (HPLC)

Synonym(s):

MLA, [1α,4(S),6β,14α,16β]-20-Ethyl-1,6,14,16-tetramethoxy-4-[[[2-(3-methyl-2,5-dioxo-1-pyrrolidinyl)benzoyl]oxy]methyl]aconitane-7,8-diol citrate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H50N2O10 · xC6H8O7
CAS Number:
Molecular Weight:
682.80 (salt-free basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

biological source

Delphinium brownii seeds

Assay

≥96% (HPLC)

color

white

solubility

H2O: 42 mg/mL

storage temp.

−20°C

SMILES string

O=C(OC[C@]12[C@@]([C@]3([C@H](CC1)OC)[C@@H]4N(C2)CC)([H])[C@@H]([C@]4(O)[C@]5(O)[C@]6([H])[C@@]3([H])C[C@@]([C@H](C5)OC)([H])[C@@H]6OC)OC)C7=CC=CC=C7N8C([C@@H](C)CC8=O)=O.OC(CC(CC(O)=O)(C(O)=O)O)=O

InChI

1S/C37H50N2O10.C6H8O7/c1-7-38-17-34(18-49-32(42)20-10-8-9-11-23(20)39-26(40)14-19(2)31(39)41)13-12-25(46-4)36-22-15-21-24(45-3)16-35(43,27(22)28(21)47-5)37(44,33(36)38)30(48-6)29(34)36;7-3(8)1-6(13,5(11)12)2-4(9)10/h8-11,19,21-22,24-25,27-30,33,43-44H,7,12-18H2,1-6H3;13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)/t19-,21+,22+,24-,25-,27+,28-,29+,30-,33?,34-,35+,36-,37+;/m0./s1

InChI key

INBLZNJHDLEWPS-DDIMIZGISA-N

Application

Methyllycaconitine citrate salt has been used as an α7 nicotinic acetylcholine receptor (α7 nAChR) antagonist:
  • to study its effects on inflammatory response in rats post nicotine treatment[1]
  • to block the activity of galantamine[2]
  • to study its effects on the hepatic branch of the vagus nerve (hVNS) in rats[3]

Biochem/physiol Actions

Methyllycaconitine (MLA) is an α7 nicotinic acetylcholine receptor (α7 nAChR) antagonist. It is a norditerpenoid alkaloid synthesized by several species of Delphinium.[4] MLA binds to the binding site of neuronal α-bungarotoxin.[5] Low doses of MLA are associated with improvement of cognition in animals.[4]

Features and Benefits

This compound is featured on the Acetylcholine Receptors (Nicotinic) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francesca Prestori et al.
PloS one, 8(5), e64828-e64828 (2013-06-07)
The brain needs mechanisms able to correlate plastic changes with local circuit activity and internal functional states. At the cerebellum input stage, uncontrolled induction of long-term potentiation or depression (LTP or LTD) between mossy fibres and granule cells can saturate
Cecília Cerqueira Café-Mendes et al.
Neuroscience letters, 636, 218-224 (2016-12-17)
The hippocampus is a brain region that is rich in nicotinic acetylcholine receptors (nAChRs), especially the α7 subtype. The hippocampus is severely affected in disorders that have a neuroinflammatory component, such as Alzheimer's disease, Parkinson's disease, and schizophrenia. Previous studies
Elizabeth Glenn Guy et al.
Psychopharmacology, 225(2), 429-440 (2012-08-14)
Stimuli associated with nicotine can become motivationally significant and may play a role in tobacco dependence. Previous work indicates that nicotine enhances responding for a conditioned reinforcer (CR). These studies examined the effects of prior exposure to nicotine on responding
Natalia Pinilla-Echeverri et al.
Circulation. Cardiovascular interventions, 13(7), e008768-e008768 (2020-07-11)
Complete revascularization with routine percutaneous coronary intervention of nonculprit lesions after primary percutaneous coronary intervention improves outcomes in ST-segment-elevation myocardial infarction. Whether this benefit is associated with nonculprit lesion vulnerability is unknown. In a prospective substudy of the COMPLETEs trial
Masaru Ishibashi et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 34(12), 2529-2547 (2009-07-25)
Identifying the neurological mechanisms underlying nicotine reinforcement is a healthcare imperative, if society is to effectively combat tobacco addiction. The majority of studies of the neurobiology of addiction have focused on dopamine (DA)-containing neurons of the ventral tegmental area (VTA).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service