M1134
L-(−)-Mannose
≥99% (GC)
Synonym(s):
L-Mannopyranose
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein:
1724628
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25
Recommended Products
Assay
≥99% (GC)
form
powder
technique(s)
gas chromatography (GC): suitable
color
beige
mp
129-131 °C (lit.)
solubility
water: 100 mg/mL, clear, colorless to very faintly yellow
SMILES string
OC[C@@H]1OC(O)[C@H](O)[C@H](O)[C@H]1O
InChI
1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5+,6?/m0/s1
InChI key
WQZGKKKJIJFFOK-JFNONXLTSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Mannose is a monosaccharide.
Application
L-Mannose has been used in a study to assess substrate specificity of galactokinase from Streptococcus pneumoniae. It has also been used in a study to investigate a versatile route to L-hexoses.
Other Notes
To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Optimized
Emily Wu et al.
In vitro cellular & developmental biology. Plant : journal of the Tissue Culture Association, 50(1), 9-18 (2014-01-01)
Annalisa Guaragna et al.
Organic letters, 8(21), 4863-4866 (2006-10-06)
[reaction: see text] An efficient route for the synthesis of orthogonally protected l-sugars has been opened up, starting from the heterocyclic homologating agent 1 and 2,3-O-isopropylidene-l-glyceraldehyde (2). Our synthetic path enables the synthesis of a 2,3-unsaturated-l-pyranoside, which can be suitably
Pin Guo et al.
Journal of experimental & clinical cancer research : CR, 39(1), 202-202 (2020-10-01)
Globally, colorectal cancer (CRC) affects more than 1 million people each year. In addition to non-modifiable and other environmental risk factors, Fusobacterium nucleatum infection has been linked to CRC recently. In this study, we explored mechanisms underlying the role of
Lars Andresen et al.
Journal of immunology (Baltimore, Md. : 1950), 188(4), 1847-1855 (2012-01-10)
NKG2D ligand surface expression is important for immune recognition of stressed and neotransformed cells. In this study, we show that surface expression of MICA/B and other NKG2D ligands is dependent on N-linked glycosylation. The inhibitor of glycolysis and N-linked glycosylation
Yang Zou et al.
Bioorganic & medicinal chemistry letters, 22(10), 3540-3543 (2012-04-20)
Galactokinases (GalKs) have attracted significant research attention for their potential applications in the enzymatic synthesis of unique sugar phosphates. The galactokinase (GalKSpe4) cloned from Streptococcus pneumoniae TIGR4 presents a remarkably broad substrate range including 14 diverse natural and unnatural sugars.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service