Skip to Content
MilliporeSigma

M0253

L-Mimosine from Koa hoale seeds

≥98%

Synonym(s):

(S)-α-Amino-β-[1-(3-hydroxy-4-oxopyridine)]propionic acid, Leucenol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C8H10N2O4
CAS Number:
Molecular Weight:
198.18
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352209
EC Number:
207-905-1
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98%

SMILES string

N[C@@H](CN1C=CC(=O)C(O)=C1)C(O)=O

InChI

1S/C8H10N2O4/c9-5(8(13)14)3-10-2-1-6(11)7(12)4-10/h1-2,4-5,12H,3,9H2,(H,13,14)/t5-/m0/s1

InChI key

WZNJWVWKTVETCG-YFKPBYRVSA-N

General description

L-mimosine [β-N (3-hydroxy-4-pyridone)-α-amino propionic acid] is a non-protein amino acid and is a vital component of tropical legumes, such as Leucaena glauca and other legumes belonging to Mimosa spp. Structurally L-mimosine resembles dihydroxyphenylalanine except 3,4-dihydroxy-phenyl ring replaced by 3-hydroxy-4-pyridone ring.

Application

L-Mimosine from Koa hoale seeds has been used to study its anti-inflammatory effect in chronic inflammatory response (potassium permanganate - induced chronic granuloma).

Biochem/physiol Actions

L-Mimosine is a plant amino acid and potential inhibitor of the cell cycle giving rise to growth arrest in G1-phase. It is an iron chelator that inhibits DNA replication in mammalian cells. L-Mimosine has been shown to promote apoptosis in xenotransplanted human pancreatic cancer. L-Mimosine stabilizes hypoxia inducible factor 1 (HIF-1) and stimulates the expression of B-cell translocation gene 2 (Btg2) and N-myc downstream regulated gene 1 (Ndrg1) at the transcriptional level, which reduce cell proliferation of prostate carcinoma cells in vitro. L-Mimosine stabilizes HIF-1 through the inhibition of prolyl hydroxylases (PHDs) which target HIF-1 through degradation. The mechanism of inhibition is likely through the chelation of Fe2+ bound to the active site of PHD which is required for enzymatic activity. Mimosine inhibits cell cycle progression via iron chelation in MDA-MB-453 human breast cancer cells. Mimosine, hinder folate metabolism in cell-specific manner.


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library