Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

LIDFA45211-EP

Lid for Eppendorf(R) FA-45-24-11 Rotor

aerosol-tight, for 5424/5424R, aluminum

Synonym(s):

Lid for Eppendorf® FA-45-48-11 Rotor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103913

material

aluminum

feature

aerosol-tight

manufacturer/tradename

Eppendorf® 022653024 (NA)
Eppendorf® 5424703003 (EU)

technique(s)

centrifugation: suitable

compatibility

for use with 5424 R
for use with 5424

Looking for similar products? Visit Product Comparison Guide

General description

Aerosol-tight QuickLock rotor lid made of aluminum. It is ideal for the FA-45-24-11 rotor.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service