Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.75
UNSPSC Code:
12352207
Quality Segment
form
liquid
usage
sufficient for 50 or 5x50 set Decontamination by wiping off surfaces (wet wipes)
packaging
pkg of 5 × 50 ea Count (in refill sachets for dispenser box), pkg of 50 ea Count Refill (in dispenser box)
bp
100 °C
density
0.999 g/mL
shipped in
ambient
storage temp.
room temp
General description
Lookout® DNA Erase Wipes are ready-to-use wet wipes for the elimination of DNA, RNA, DNases, and RNases from a variety of surfaces and are particularly suitable for decontamination of PCR workstations, electrical devices, pipetting equipment.
This product represents a great alternative to standard, inefficient cleaning methods (e.g. with sponges or paper towels), which might help spread contaminations rather than prevent them.
All most common contaminants (DNA, RNA, DNases, and RNases) are effectively removed at once from the contaminated surface within seconds after use.
The wipes are pre-soaked with a solution contains surfactants and a non-alkaline and non-carcinogenic agent.
This product represents a great alternative to standard, inefficient cleaning methods (e.g. with sponges or paper towels), which might help spread contaminations rather than prevent them.
All most common contaminants (DNA, RNA, DNases, and RNases) are effectively removed at once from the contaminated surface within seconds after use.
The wipes are pre-soaked with a solution contains surfactants and a non-alkaline and non-carcinogenic agent.
Application
Recommended Use:
Applicable in research and industry, only. Not recommended for clinical applications. Use as directed. DNA Erase Wipes can be used on glass, ceramic, plastic, rubber, steel, and precious metal. Cleaning of light (e.g. aluminum) or non-ferrous metals with DNA Erase Wipes is not recommended. Always test a small area first to make sure the product does not cause damage or discoloration (e.g. on coated or sensitive materials).
Applicable in research and industry, only. Not recommended for clinical applications. Use as directed. DNA Erase Wipes can be used on glass, ceramic, plastic, rubber, steel, and precious metal. Cleaning of light (e.g. aluminum) or non-ferrous metals with DNA Erase Wipes is not recommended. Always test a small area first to make sure the product does not cause damage or discoloration (e.g. on coated or sensitive materials).
Preparation Note
When stored at room temperature, the product is stable until the expiry date indicated on the label.
Other Notes
No Consumables and Lab Devices are required.
Legal Information
LookOut is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.