Skip to Content
MilliporeSigma

L9042

LookOut® DNA Erase

Refill

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352207
NACRES:
NA.75
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

liquid

storage condition

dry at room temperature

technique(s)

DNA purification: suitable

application(s)

microbiology

shipped in

ambient

storage temp.

room temp

Application

Both the 250 ml spray reagent (Product Number L8917) and the 1 L refill (Product Number L9042) are supplied ready-to-use.
LookOut™ DNA Erase is a potent, ready-to-use solution for rapid DNA decontamination of surfaces in laboratories. This reagent is characterized by its high efficiency. The decontamination spray completely destroys DNA within 60 seconds of surface treatment. The solution contains a unique combination of DNA destroying and surface active agents.

Biochem/physiol Actions

LookOut DNA Erase may be applied to steel, glass, ceramic, plastic, rubber, or precious metals including laboratory equipment and pipettors. It should not be applied on light and non-ferrous metals. For other sensitive surfaces, test in a small area before applying.

Legal Information

LookOut is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under LookOut® DNA Erase


pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

flash_point_f

Not applicable

flash_point_c

Not applicable

Storage Class

12 - Non Combustible Liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library