Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L9026

Sigma-Aldrich

Lys-Lys-Lys-Lys

≥95% (TLC)

Synonym(s):

Lys4, Tetralysine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H50N8O5
CAS Number:
Molecular Weight:
530.70
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

Lys-Lys-Lys-Lys, ≥95% (TLC)

Quality Level

Assay

≥95% (TLC)

form

powder or crystals

composition

Peptide content, ≥50%

color

white

storage temp.

−20°C

SMILES string

NCCCCC(N)C(=O)NC(CCCCN)C(=O)NC(CCCCN)C(=O)NC(CCCCN)C(O)=O

InChI

1S/C24H50N8O5/c25-13-5-1-9-17(29)21(33)30-18(10-2-6-14-26)22(34)31-19(11-3-7-15-27)23(35)32-20(24(36)37)12-4-8-16-28/h17-20H,1-16,25-29H2,(H,30,33)(H,31,34)(H,32,35)(H,36,37)

InChI key

RRBGTUQJDFBWNN-UHFFFAOYSA-N

Amino Acid Sequence

Lys-Lys-Lys-Lys

Biochem/physiol Actions

Short poly-L-lysines polypeptides such as trilysine (lys3), tetralysine (tetra-L-lysine, lys4) and pentalysine (penta-L-lysine, lys5) are cationic moieties that may be used in the construction of gene delivery vectors and DNA nanoparticles.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maja Semanjski et al.
Science signaling, 11(547) (2018-09-13)
The bacterial serine-threonine protein kinase HipA promotes multidrug tolerance by phosphorylating the glutamate-tRNA ligase (GltX), leading to a halt in translation, inhibition of growth, and induction of a physiologically dormant state (persistence). The HipA variant HipA7 substantially increases persistence despite
Tohru Fujiwara et al.
Biochemical and biophysical research communications, 485(2), 380-387 (2017-02-22)
The transcription factor GATA-1-interacting protein Friend of GATA-1 (FOG1) is essential for proper transcriptional activation and repression of GATA-1 target genes; yet, the mechanisms by which FOG1 exerts its activating and repressing functions remain unknown. Forced FOG1 expression in human
Donghong Zhang et al.
Nature communications, 8(1), 1979-1979 (2017-12-08)
Despite the importance of cardiomyocyte proliferation in cardiac development and regeneration, the mechanisms that promote cardiomyocyte cell cycle remain incompletely understood. RE1 silencing transcription factor (REST) is a transcriptional repressor of neuronal genes. Here we show that REST also regulates
Adele Aronica et al.
Biochimica et biophysica acta, 1861(1 Pt A), 3210-3220 (2016-08-19)
CA19.9 antigen has been assumed as an abundant product of cancer cells, due to the reactivity found by immunohistochemical staining of cancer tissues with anti-CA19.9 antibody. Expression and biosynthesis of type 1 chain Lewis antigens in the colon and the
Anne L Jaquiery et al.
Pediatric research, 80(6), 861-869 (2016-08-17)
The nutritional plane and composition during fetal life can impact upon growth and epigenetic regulation of genes affecting pancreatic β-cell development and function. However, it is not clear whether β-cell development can be altered by nutritional factors or growth rate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service