Skip to Content
MilliporeSigma

L8384

Linoleic Acid-Albumin from bovine serum albumin

γ-irradiated, lyophilized powder, BioReagent, suitable for cell culture

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.75
UNSPSC Code:
12352202
Biological source:
bovine plasma
Form:
lyophilized powder
Technique(s):
cell culture | mammalian: suitable
Impurities:
≤50 EU/mg endotoxin
Concentration:
>0.5% (linolenic acid)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine plasma

Quality Segment

description

Contains 1.1-2.3 moles linoleic acid per mole BSA

sterility

γ-irradiated

product line

BioReagent

form

lyophilized powder

concentration

>0.5% (linolenic acid)

technique(s)

cell culture | mammalian: suitable

impurities

≤50 EU/mg endotoxin

solubility

water: 10 mg/mL, clear to hazy, colorless to light yellow

storage temp.

2-8°C

General description

Linoleic acid is an essential and highly consumed polyunsaturated fatty acid (PUFA). Albumin is a globular protein. Bovine serum albumin (BSA) is composed of a single polypeptide chain and cross-linked cysteine disulfide bridges. BSA is heart-shaped and is made up of three homologous domains I-III and each domain has two subdomains A and B.

Application

Linoleic Acid-Albumin from bovine serum albumin has been used: in cell culture and to study its effects on the human colonic adenocarcinoma cell line. It has also been used in the in vitro culture (IVC) of cow embryos/zygotes. The binding of linoleic acid to BSA improves its stability and solubility in culture media.

Biochem/physiol Actions

Linoleic acid plays a role in maintaining the transdermal water barrier in epidermis. Serum albumin constitutes about 52-62% of the total water-soluble fraction proteins in the blood plasma.


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library