Skip to Content
MilliporeSigma

L5295

LY 320135

≥98% (HPLC)

Synonym(s):

4-[6-Methoxy-2-(4-methoxyphenyl)benzofuran-3-carbonyl]benzonitrile, LY320135, [6-Methoxy-2-(4-methoxyphenyl)benzo[b]furan-3-yl](4-cyanophenyl)methanone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C24H17NO4
CAS Number:
Molecular Weight:
383.40
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

color

orange

solubility

DMSO: ≥10 mg/mL

originator

Eli Lilly

storage temp.

2-8°C

SMILES string

COc1ccc(cc1)-c2oc3cc(OC)ccc3c2C(=O)c4ccc(cc4)C#N

InChI

1S/C24H17NO4/c1-27-18-9-7-17(8-10-18)24-22(20-12-11-19(28-2)13-21(20)29-24)23(26)16-5-3-15(14-25)4-6-16/h3-13H,1-2H3

InChI key

RYNSGDFWBJWWSZ-UHFFFAOYSA-N

Biochem/physiol Actions

LY320135 is a potent CB1 receptor antagonist/inverse agonist (Ki = 141 nM) with greater than 70-fold selectivity over CB2 receptors (Ki > 10 μM).
LY320135 is a potent CB1 receptor antagonist/inverse agonist (Ki = 141 nM) with greater than 70-fold selectivity over CB2 receptors (Ki > 10 μM). Structurally dissimilar from SR 141716A and AM 251. Shows weak binding to both 5-HT2 (Ki = 6.4 μM) and muscarinic receptors (Ki = 2.1 μM)

Features and Benefits

This compound is featured on the Cannabinoid Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Eli Lilly. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library