Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

L4793

Sigma-Aldrich

Anti-LSD1 (AOF2) (N-terminal) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-Amine oxidase, flavin-containing, 2, Anti-BHC110, Anti-BRAF-HDAC complex protein 110, Anti-KIAA0601 protein, Anti-Lysine-specific demethylase 1, Anti-NPAO, Anti-Nuclear Polyamine Oxidase

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 110-115 kDa (two bands)

species reactivity

mouse (predicted), rat (predicted), human

technique(s)

immunoprecipitation (IP): 2-5 μL using MCF7 cell lysates
microarray: suitable
western blot: 1:1,000-1:2,000 using MCF7 cell lysates (2 bands, 2 isoforms)

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KDM1A(23028)
mouse ... KDM1A(99982)
rat ... KDM1A(500569)

General description

Lysine specific demethylase 1 (LSD1) also called Amine oxidase flavin-containing domain 2 (AOF2) is a nuclear protein containing a Swi3p, Rsc8p and Moira (SWIRM) domain, a flavin adenine dinucleotide (FAD)-binding motif, and an amine oxidase domain. LSD1 gene is mapped to human chromosome 1p36.12.

Specificity

Anti-LSD1 (AOF2)(N-terminal) specifically recognizes human LSD1 (AOF2).

Immunogen

synthetic peptide corresponding to amino acids 123-139 of human LSD1 (AOF2) (isoform b), conjugated to KLH via a C-terminal added cysteine residue. The immunizing sequence is present also in isoform a. This sequence is conserved in human, rat, and mouse.

Application

Anti-LSD1 (AOF2) (N-terminal) antibody produced in rabbit has been used in immunoblotting and immunoprecipitation.

Biochem/physiol Actions

Lysine specific demethylase 1 (LSD1) is a component of several histone deacetylase complexes that function through modifying chromatin structure to repress transcription. Methylation of lysine can lead either to gene silencing or activation depending on the specific lysine residue that is methylated. LSD1 action can signal either activation or repression of transcription. The demethylation of histone H3 Lys4 by LSD1 leads to transcriptional repression of target genes, while demethylation of histone H3 at Lys9 leads to de-repression of androgen receptor target genes. The activity of LSD1 is regulated by its associated factors. Corepressor of RE1 silencing transcription factor/neural-restrictive silencing factor (REST), a transcriptional corepressor, promotes demethylation by enhancing the association of LSD1 with the nucleosomes and also protects LSD1 from proteasomal degradation.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seung Eun Yu et al.
Biochemical and biophysical research communications, 427(2), 336-342 (2012-09-25)
S100A7, a member of S100 calcium binding protein family, is highly associated with breast cancer. However, the molecular mechanism of S100A7 regulation remains unclear. Here we show that long-term treatment with estradiol stimulated S100A7 expression in MCF7 breast cancer cells
Ruth Ramírez-Ramírez et al.
Journal of clinical pathology, 73(2), 107-111 (2019-09-01)
KDM1A/LSD1 and ZNF217 are involved in a protein complex that participates in transcriptional regulation. ZNF217 has been analysed in numerous cancers and its amplification has been associated with advanced stages of disease; however, a similar role for KDM1A/LSD1 has not
Yu-Jiang Shi et al.
Molecular cell, 19(6), 857-864 (2005-09-06)
LSD1 is a recently identified human lysine (K)-specific histone demethylase. LSD1 is associated with HDAC1/2; CoREST, a SANT domain-containing corepressor; and BHC80, a PHD domain-containing protein, among others. We show that CoREST endows LSD1 with the ability to demethylate nucleosomal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service