Skip to Content
MilliporeSigma

L4250

Dodecanoic acid

≥99% (GC/titration)

Synonym(s):

Lauric acid, NSC 5026

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3(CH2)10COOH
CAS Number:
Molecular Weight:
200.32
UNSPSC Code:
12161501
NACRES:
NA.77
PubChem Substance ID:
EC Number:
205-582-1
Beilstein/REAXYS Number:
1099477
MDL number:
Assay:
≥99% (GC/titration)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥99% (GC/titration)

SMILES string

CCCCCCCCCCCC(O)=O

InChI

1S/C12H24O2/c1-2-3-4-5-6-7-8-9-10-11-12(13)14/h2-11H2,1H3,(H,13,14)

InChI key

POULHZVOKOAJMA-UHFFFAOYSA-N

Application


  • Biopolymer synthesis for agricultural use: A study engineered Pseudomonas chlororaphis to synthesize biopolymers incorporating 3-hydroxybutyrate and medium-chain-length 3-hydroxyalkanoates from dodecanoic acid. This process highlights its role in sustainable bioplastic production, which could benefit agricultural practices by developing biodegradable mulch films (Deng et al., 2024).

Biochem/physiol Actions

Substrate for CYP 4A11


Still not finding the right product?

Explore all of our products under Dodecanoic acid


pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

348.8 °F - closed cup

flash_point_c

176 °C - closed cup

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library