Skip to Content
MilliporeSigma

L3536

Greiner multiwell plate lids

for standard multiwell plate, clear, sterile

Synonym(s):

microplate lids, microtiter plate lids, multiwell plate lids, Greiner 656161, microplate cover

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.22
UNSPSC Code:
41122107
Material:
colorless wells, polystyrene
Size:
96 wells
Sterility:
sterile
Binding type:
Tissue Culture (TC)-treated surface
Feature:
lid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

colorless wells, polystyrene

sterility

sterile

feature

lid

packaging

case of 100

manufacturer/tradename

Greiner 656161

size

96 wells

color

clear

binding type

Tissue Culture (TC)-treated surface

General description

Corner notches for proper orientation. Robotics friendly full- and Lo-profile skirt options. RNase and DNase free.
Microplate Lid, Sterile, Single Position, High Profile, without Condensation Rings

Application

Sterile Lid for Polystyrene MicroPlates, DNAse/RNAse, Human DNA and Pyrogen free.

Features and Benefits

  • Greiner multiwell plate lids are made of polystyrene.
  • Corner notches for proper orientation.
  • Robotics-friendly full- and Lo-profile skirt options.
  • Free of detectable RNase and DNase.

Other Notes

length: 127,5 mm; width: 85 mm; curvature approx. 300 μM. Lid height 9 mm

Legal Information



Still not finding the right product?

Explore all of our products under Greiner multiwell plate lids




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library