Skip to Content
MilliporeSigma
All Photos(2)

Documents

L0756

Sigma-Aldrich

Lung acetone powder from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.25

biological source

rabbit

form

powder

storage temp.

−20°C

Application

Lung acetone powder from rabbit has been used in the determination of angiotensin converting enzyme (ACE) inhibitory activity:
  • from milk fermentate by Bifidobacterium longum
  • in Caco-2 brush border expressing (Bbe) cell monolayer
  • from ginger extracts

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spectrophotometric assay and properties of the angiotensin-converting enzyme of rabbit lung.
D W Cushman et al.
Biochemical pharmacology, 20(7), 1637-1648 (1971-07-01)
Physiological activities of ginger extracts originated from different habitat
Food Science and Biotechnology, 15(1), 143-147 (2006)
V Vermeirssen et al.
Journal of peptide science : an official publication of the European Peptide Society, 8(3), 95-100 (2002-04-05)
ACE inhibitory peptides are biologically active peptides that play a role in blood pressure regulation. When derived from food proteins during food processing or gastrointestinal digestion, these peptides could function as efficient agents in treating and preventing hypertension. However, in
Go Eun Ha et al.
Korean journal for food science of animal resources, 35(6), 738-747 (2016-02-16)
Angiotensin-converting enzyme (ACE) inhibitory activity was evaluated for the low-molecular-weight fraction (<3 kDa) obtained from milk fermentation by Bifidobacterium longum KACC91563. The ACE inhibitory activity in this fraction was 62.3%. The peptides generated from the <3 kDa fraction were identified

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service