Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

K3628

Sigma-Aldrich

Kininogen, low molecular weight from human plasma

≥95% (SDS-PAGE), lyophilized powder

Synonym(s):

LMW kininogen

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human plasma

Quality Level

Assay

≥95% (SDS-PAGE)

form

lyophilized powder

mol wt

64000 Da

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

Application

Kininogen, low molecular weight from human plasma has been used to study rMASP-1 (recombinant mannose-binding lectin-associated serine protease 1)-mediated cleavage of LK (low molecular weight kininogen). It has been used as a standard for protein identification in milk-proteomics study.

Biochem/physiol Actions

LMW kininogen is a major extracellular cysteine proteinase inhibitor. Tissue kallikrein releases bradykinin, a potent bronchoconstrictor and vasoactive agent, from both HMW and LMY kininogen.

Physical form

Lyophilized from a solution in sodium acetate and sodium chloride.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Delphine Vincent et al.
Frontiers in genetics, 6, 360-360 (2016-01-23)
Milk is a complex fluid whose proteome displays a diverse set of proteins of high abundance such as caseins and medium to low abundance whey proteins such as ß-lactoglobulin, lactoferrin, immunoglobulins, glycoproteins, peptide hormones, and enzymes. A sample preparation method
József Dobó et al.
PloS one, 6(5), e20036-e20036 (2011-06-01)
Bradykinin (BK), generated from high-molecular-weight kininogen (HK) is the major mediator of swelling attacks in hereditary angioedema (HAE), a disease associated with C1-inhibitor deficiency. Plasma kallikrein, activated by factor XIIa, is responsible for most of HK cleavage. However other proteases
Xavier Charest-Morin et al.
Pharmacology research & perspectives, 3(2), e00119-e00119 (2015-06-04)
Tissue kallikrein (KLK-1), a serine protease, initiates the release of bradykinin (BK)-related peptides from low-molecular weight kininogen. KLK-1 and the BK B2 receptor (B2R) mediate beneficial effects on the progression of type 2 diabetes and renal disease, but the precise

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service