Skip to Content
MilliporeSigma

K1876

Kanamycin disulfate salt from Streptomyces kanamyceticus

aminoglycoside antibiotic

Synonym(s):

Kanamycin Disulfate Salt, Kanamycin disulfate, O-3-amino-3-deoxy-alpha-D-glucopyranosyl-(1->6)-O-(6-amino-6-deoxy-alpha-D-glucopyranosyl-(1->4))-2-deoxy-D-Streptamine sulfate (1:2) (salt), Kanamycin A, Kanamycin acid sulfate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
NACRES:
NA.76
PubChem Substance ID:
UNSPSC Code:
51281654
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

Streptomyces kanamyceticus

Quality Segment

form

powder

potency

~650 μg per mg

impurities

≤4% Kanamycin B

color

white to off-white

solubility

H2O: 10 mg/mL

antibiotic activity spectrum

Gram-negative bacteria, Gram-positive bacteria, mycobacteria, mycoplasma

mode of action

protein synthesis | interferes

SMILES string

OS(O)(=O)=O.OS(O)(=O)=O.NC[C@H]1O[C@H](O[C@@H]2[C@@H](N)C[C@@H](N)[C@H](O[C@H]3O[C@H](CO)[C@@H](O)[C@H](N)[C@H]3O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C18H36N4O11.2H2O4S/c19-2-6-10(25)12(27)13(28)18(30-6)33-16-5(21)1-4(20)15(14(16)29)32-17-11(26)8(22)9(24)7(3-23)31-17;2*1-5(2,3)4/h4-18,23-29H,1-3,19-22H2;2*(H2,1,2,3,4)/t4-,5+,6-,7-,8+,9-,10-,11-,12+,13-,14-,15+,16-,17-,18-;;/m1../s1

InChI key

OGTKIXVMLDAMNU-KNQICTBBSA-N

General description

Chemical structure: aminoglycoside

Application

Kanamycin sulfate is a broad spectrum minoglycoside-antibiotic It is used as an additive in culture media for the isolation of group D Streptococci on Kanamycin Esculin Azide Agar (Mossel et al.) or for selection of transformed plant cells containing the neomycin phosphotransferase (for kanamycin resistance); on a kanamycin-medium. Kanamycin has been used for electrophysiological recordings of outer hair cells and inner hair cells. It is used to inhibit shoot regeneration from Siberian Elmleaf explants. It is recommended for use in cell culture applications at 100 mg/mL.

Biochem/physiol Actions

Mode of Action: The product acts by binding to the 70S ribosomal subunit, inhibiting translocation and eliciting miscoding.

Mode of Resistance: Aminoglycoside-modifying enzymes (including acetyltransferase, phosphotransferase, nucleotidyltransferase) can alter this antibiotic, preventing its interaction with ribosomes.

Antimicrobial Spectrum: Kanamycin sulfate is effective against gram-negative and gram-postiive bacteria, and mycoplasma.

Preparation Note

Kanamycin acid sulfate is prepared by solubilizing kanamycin sulfate in dilute sulfuric acid and drying. The sulfate content is approximately 25%.
Kanamycin sulfate is soluble in water at 10 mg/mL, yielding a clear solution. It is practically insoluble in alcohol, acetone, chloroform, ether and ethyl acetate. A 1% solution in water has a pH of 6.5 to 8.5. Sterile solutions can be prepared by a sterile filtration, through a .2μm filter.

Disclaimer

Solutions are stable at 37°C for approximately 5 days. Aqueous stock solutions can be stored at 2-8°C for long term storage.


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library