Skip to Content
MilliporeSigma

I9659

Monoclonal Anti-Insulin-like Growth Factor-II antibody produced in mouse

clone 75014, purified immunoglobulin, lyophilized powder

Synonym(s):

Anti-IGF2, Anti-INSIGF

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
51111800
NACRES:
NA.41
MDL number:
Conjugate:
unconjugated
Clone:
75014, monoclonal
Application:
WB
Citations:
4
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

75014, monoclonal

form

lyophilized powder

species reactivity

human

technique(s)

western blot: 1-2 μg/mL

isotype

IgG2a

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... IGF2(3481)

Immunogen

purified, E. coli-derived recombinant human insulin-like growth factor-II.

Biochem/physiol Actions

Insulin-like growth factors -I and -II are important regulators of signaling that mediates growth and metabolism in cells. Insulin-like growth factor -II (IGF-II) is widely expressed during embryonic stages and regulates the placental development. IGF-II acts via specific receptor, IGF-IIR, the binding does not result in signal transduction, but is a mechanism to restrict the bioavailability of IGF-II. The effects of IGF-II signaling are important in growth of blood vessels (angiogenesis) gene transcription, proliferation and transformation.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


Storage Class

13 - Non Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library