Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I9154

Sigma-Aldrich

Interleukin-10 from rat

>95% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder

Synonym(s):

IL-10

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

rat

Quality Level

recombinant

expressed in E. coli

Assay

>95% (SDS-PAGE)

form

lyophilized powder

potency

1-35 ng/mL ED50

mol wt

predicted mol wt ~19 kDa

packaging

pkg of 5 μg

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

rat ... Il10(25325)

Application

Interleukin-10 (IL-10) from rat has been used to study the anti-allodynic effect of IL-10 in rat model.

Biochem/physiol Actions

Interleukin-10 is an important regulator of the functions of lymphoid and myeloid cells. IL-10 can block the activation of cytokine synthesis and several accessory functions of macrophages. Human and mouse IL-10 share a 73% sequence homology. However, human IL-10 acts on both human and mouse target cells, while mouse IL-10 has species-specific activity. In mouse, the cellular sources of IL-10 consist of Th2 cells, activated fetal thymocytes, macrophages, keratinocytes, LY-1+ (CD5+), and normal B cells. In human, the cellular sources of IL-10 consist of CD4+ T cells and T cell clones, thymocytes, B cells, B cell lymphomas, macrophages, mast cell lines and keratinocytes. IL-10 stimulates the growth of stem cells, mast cells and thymocytes. IL-10 enhances cytotoxic T cell development, and co-stimulates B cell differentiation and Ig secretion. IL-10 regulates angiogenesis by inducing the cell-type dependent expression of either angiogenic or angiostatic factors.
Interleukin-10 regulates lymphoid and myeloid cell functions. It blocks the activation of cytokine synthesis and several accessory functions of macrophages. IL-10 enhances cytotoxic T cell development and co-stimulates B cell differentiation and Ig secretion. IL-10 regulates angiogenesis by inducing cell-type dependent expression of either angiogenic or angiostatic factors.
Interleukin-10 regulates lymphoid and myeloid cell functions; blocks the activation of cytokine synthesis and several accessory functions of macrophages; enhances cytotoxic T cell development and co-stimulates B cell differentiation and Ig secretion.

Physical form

Lyophilized from a 0.2 μm filtered in 20 mM Tris, 0.1 M sodium chloride containing 50 μg of bovine serum albumin per 1 μg of cytokine.

Analysis Note

The biological activity is measured in a cell proliferation assay using the murine mast cell line, MC/9.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Byung-Sang Lee et al.
Journal of Korean medical science, 28(2), 308-314 (2013-02-13)
We examined the possible anti-inflammatory mechanisms of gabapentin in the attenuation of neuropathic pain and the interaction between the anti-allodynic effects of gabapentin and interleukin-10 (IL-10) expression in a rat model of neuropathic pain. The anti-allodynic effect of intrathecal gabapentin
Yoshiyuki Goto et al.
Scientific reports, 5, 15918-15918 (2015-11-03)
Fucosylated glycans on the surface of epithelial cells (ECs) regulate intestinal homeostasis by serving as attachment receptors and a nutrient source for some species of bacteria. We show here that epithelial fucosylation in the ileum is negatively regulated by IL-10-producing
Justine Durand et al.
Journal of immunology (Baltimore, Md. : 1950), 195(10), 5035-5044 (2015-10-04)
Emerging knowledge regarding B cells in organ transplantation has demonstrated that these cells can no longer be taken as mere generators of deleterious Abs but can also act as beneficial players. We previously demonstrated in a rat model of cardiac
Piyatida Tangteerawatana et al.
The Southeast Asian journal of tropical medicine and public health, 45(3), 517-530 (2014-07-01)
Immunity to malaria can be acquired but only after repeat exposures to polymorphic Plasmodium. However, the development of clinical outcomes during P. falciparum infection is not clearly understood. This study elucidated whether monocytes, neutrophils and pro/anti-inflammatory cytokines were associated with
Mohamad Bouhamdan et al.
Cellular signalling, 27(10), 2068-2076 (2015-07-26)
The mitogen activated protein kinases ERK1/2 play an important role in response to toll like receptor (TLR) activation and cytokine production, including IL-10 and IL-12. Here, we examined the role of MEK1 in ERK1/2 activation in response to TLR4 agonist

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service