Skip to Content
MilliporeSigma

Skip To

I8904

Insulin-like Growth Factor-II from mouse

>97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder

Synonym(s):

IGF-II

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77
MDL number:
Biological source:
mouse
Recombinant:
expressed in E. coli
Assay:
>97% (SDS-PAGE)
Form:
lyophilized powder
Mol wt:
~7.4 kDa
Impurities:
<1.0EUper1mg (LAL test)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Insulin-like Growth Factor-II from mouse, IGF-II, recombinant, expressed in E. coli, lyophilized powder

biological source

mouse

recombinant

expressed in E. coli

assay

>97% (SDS-PAGE)

form

lyophilized powder

quality

endotoxin tested

mol wt

~7.4 kDa

packaging

pkg of 5X50 μg, pkg of 50 μg

impurities

<1.0EUper1mg (LAL test)

UniProt accession no.

storage temp.

−20°C

Gene Information

General description

Recombinant Mouse IGF-II is produced from a DNA sequence encoding the mature IGF-II protein. Mouse IGF-II, a 67 amino acid protein, has a predicted molecular matrix of ~7.4 kDa. Mouse and human IGF-II share 91% sequence homology. Insulin-like growth factor II (also known as multiplication stimulating activity or MSA) and insulin-like growth factor I (IGF-I) belong to the family of insulin-like growth factors, which are structurally homologous to proinsulin. Mature IGF-I and IGF-II are highly conserved and share ~70% amino acid sequence identity. They have autocrine, paracrine, and endocrine functions.

Biochem/physiol Actions

Insulin-like growth factor II (IGF-II) is a potent mitogenic growth factor that mediates growth-promoting activities in embryonic development. IGF-II binds the IGF-II receptor with high affinity. IGF-I and IGF II are expressed in many tissues and cell types. IGF-II is mitogenic for a variety of cultured cells including human or chicken fibroblasts, mouse 3T3 cells, normal rat kidney.

Physical form

Lyophilized from a 0.2 μm filtered solution of 30% acetonitrile and 0.1% of TFA.

Analysis Note

The biological activity is measured in a serum-free cell proliferation assay using the human breast carcinoma cell line MCF-7.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
I3657I2526I3769
biological source

mouse

biological source

mouse

biological source

human

biological source

human

recombinant

expressed in E. coli

recombinant

expressed in NSO cells

recombinant

expressed in E. coli

recombinant

expressed in E. coli

assay

>97% (SDS-PAGE)

assay

≥95% (SDS-PAGE)

assay

≥95% (SDS-PAGE)

assay

≥95% (HPLC)

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

200

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

impurities

<1.0EUper1mg (LAL test)

impurities

endotoxin, tested

impurities

≤0.1 EU/μg (LAL test)

impurities

≤1.00 EU/μg


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters