Skip to Content
MilliporeSigma

I8885

Anti-Human IgG (Fc specific) antibody produced in goat

whole antiserum

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
MDL number:
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ELISA (i), QPA
Citations:
4
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

goat

Quality Segment

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

secondary antibodies

clone

polyclonal

contains

15 mM sodium azide

technique(s)

indirect ELISA: 1:50,000, quantitative precipitin assay: 4.0-7.0 mg/mL

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Immunoglobulin G (IgG), a widely expressed serum antibody, has two heavy chains and two light chains connected by a disulfide bond. It is a glycoprotein, usually found as a monomer. IgG is further subdivided into four classes namely, IgG1, IgG2, IgG3, and IgG4. IgG antibody subtype is the most abundant of serum immunoglobulins of the immune system. About 70 % of the total immunoglobulin consists of IgG.

Application

Anti-Human IgG (Fc specific) antibody produced in goat has been used in enzyme-linked immunosorbent assay (ELISA).

Biochem/physiol Actions

Immunoglobulin G (IgG) mainly helps in immune defense. It participates in hypersensitivity type II and type III. It provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defense of the neonate against infections.
Specificity for the Fc fragment of human IgG is determined by immunoelectrophoresis (IEP) versus normal human serum and Fc fragment of human IgG. No reactivity is observed versus purified human IgA, IgG (Fab fragment), IgM, Bence Jones kappa and Bence Jones lambda myeloma proteins.

Physical form

Goat Anti-Human IgG is supplied as a liquid containing 0.1% sodium azide (see MSDS) as a preservative.

Preparation Note

For continuous use, store at 2-8°C for up to one month. For extended storage, the solution may be frozen in working aliquots. Repeated freezing and thawing is not recommended. Storage in "frost-free" freezers is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. This goat antiserum was maintained at pH 5.0 for 40 minutes to meet U.S.D.A. requirements.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library