Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I7776

Sigma-Aldrich

Anti-Interleukin-4 antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-IL-4

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

mouse

technique(s)

neutralization: suitable
western blot: suitable

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Il4(16189)

Looking for similar products? Visit Product Comparison Guide

General description

The antibody will neutralize the biological activity of recombinant mouse IL-4, but not recombinant human IL-4. It shows <5% cross-reactivity with recombinant rat IL-4, and shows no cross-reactivity with recombinant human IL-4.

Immunogen

recombinant mouse IL-4.

Biochem/physiol Actions

Interleukin-4 (IL-4) is a T cell derived chemokine that stimulates the Th2 mediated immune response and proliferation of B cells. The effects of IL-4 are mediated by two types of receptors, type I receptor consisting of IL-4Rα and common γ chain and type II receptor composed of IL-4Rα and IL-13Rα1. The signaling stimulated by IL-4 leads to activation of JAK/STAT6 and IRS-mediated PI3K/Akt pathway. IL-4 is responsible for endocytic activity of macrophages, chemotaxis of leukocytes in response to inflammation, angiogenesis and regulation of nitric oxide metabolism in macrophages.

Physical form

Lyophilized from 0.2 μm filtered solution in phosphate buffered saline..

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hao-Wei Wang et al.
Cell cycle (Georgetown, Tex.), 9(24), 4824-4835 (2010-12-15)
Although macrophages were originally recognized as major immune effector cells, it is now appreciated that they also play many important roles in the maintenance of tissue homeostasis, and are involved in a variety of pathological conditions including cancer. Several studies
Jane Gilmour et al.
Immunology, 124(4), 437-444 (2008-05-16)
The mechanism of differentiation of naïve T cells to a variety of effector lineages, but particularly to T helper type 1 (Th1) and Th2 cells, has been the subject of intense scrutiny over the past two decades. Studies have revealed
H Jiang et al.
The Journal of allergy and clinical immunology, 105(6 Pt 1), 1063-1070 (2000-06-16)
In the past several years, extensive studies on the mechanisms underlying IL-4 and IL-13 signaling have enabled us to gain insight into how these cytokines regulate immune responses. Because both IL-4 and IL-13 use the IL-4Ralpha as a receptor component

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service