Skip to Content
MilliporeSigma

I7375

Iodonitrotetrazolium violet-formazan

crystalline

Synonym(s):

1-(4-Iodophenyl)-5-(4-nitrophenyl)-3-phenylformazan, INT-Formazan, Iodonitrotetrazolium formazan

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C19H14IN5O2
CAS Number:
Molecular Weight:
471.25
UNSPSC Code:
12352204
NACRES:
NA.83
PubChem Substance ID:
EC Number:
231-950-6
Beilstein/REAXYS Number:
1829738
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

crystalline

Quality Segment

color

brown

solubility

DMF: 50 mg/mL

storage temp.

2-8°C

SMILES string

[O-][N+](=O)c1ccc(NN=C(N=Nc2ccc(I)cc2)c3ccccc3)cc1

InChI

1S/C19H14IN5O2/c20-15-6-8-16(9-7-15)21-23-19(14-4-2-1-3-5-14)24-22-17-10-12-18(13-11-17)25(26)27/h1-13,22H/b23-21?,24-19-

InChI key

FVFWUNAWQMROIF-CDNKMLFNSA-N

Application


  • Starvation-survival processes of a marine Vibrio.: This research explores the use of Iodonitrotetrazolium Violet-Formazan for studying the starvation-survival mechanisms in marine Vibrio, highlighting its application as a metabolic activity indicator under nutrient-deprived conditions, which is crucial for understanding bacterial survival strategies in natural aquatic environments (Amy et al., 1983).



pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library