Skip to Content
MilliporeSigma
All Photos(1)

Documents

I7033

Sigma-Aldrich

6′-Iodoresiniferatoxin

amorphous semi-solid

Synonym(s):

6-I-RTX, Resiniferonyl 20-(4-hydroxy-2-iodo-5-methoxyphenylacetate 9,13,14-ortho(phenylacetate)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H39O9I
Molecular Weight:
754.60
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

amorphous semi-solid

Quality Level

color

colorless to white

storage temp.

−20°C

SMILES string

COc1cc(CC(=O)OCC2=CC3C4O[C@]6(Cc5ccccc5)O[C@]4(C[C@@H](C)[C@]3(O6)C7C=C(C)C(=O)[C@@]7(O)C2)C(C)=C)c(I)cc1O

InChI

1S/C37H39IO9/c1-20(2)35-16-22(4)37-26(33(35)45-36(46-35,47-37)18-23-9-7-6-8-10-23)12-24(17-34(42)30(37)11-21(3)32(34)41)19-44-31(40)14-25-13-29(43-5)28(39)15-27(25)38/h6-13,15,22,26,30,33,39,42H,1,14,16-19H2,2-5H3/t22-,26?,30-,33-,34-,35-,36-,37-/m1/s1

InChI key

IITCVPTZKLXSKQ-DCLOTLOASA-N

Biochem/physiol Actions

VR1 vanilloid receptor partial agonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mark E McDonnell et al.
Bioorganic & medicinal chemistry letters, 12(8), 1189-1192 (2002-04-06)
Using a 'directed' iodination procedure, novel iodo-resiniferatoxin congeners were synthesized from 4-acetoxy-3-methoxyphenylacetic acid and resiniferinol- 9,13,14-ortho-phenylacetate (ROPA). The 2-iodo-4-hydroxy-5-methoxyphenylacetic acid ester of resiniferinol 5 displayed high affinity binding (K(i)=0.71 nM) for the human vanilloid VR1 receptor and functioned as a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service