Skip to Content
MilliporeSigma

I4640

Human IgG4 Kappa, Recombinant Protein

purified immunoglobulin, buffered aqueous solution

Synonym(s):

Human IgG4 Protein

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Form:
buffered aqueous solution
Assay:
>95% (SDS-PAGE)
Biological source:
human
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

assay

>95% (SDS-PAGE)

form

buffered aqueous solution

shipped in

dry ice

storage temp.

−20°C

General description

IgG antibody subtype is the most abundant serum immunoglobulin of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to the fetus through the placenta and is vital for immune defense of the neonate against infections. IgG4, κ recombinant protein is obtained by expression of a phage library in a human cell line and purified by affinity chromatography.

Application

The purified IgG4, κ may be used as an immunoglobulin calibrator, reference antigen, blocking agent, or coating protein in a variety of immunoassays including ELISA, dot blot immunoblotting, Western immunoblotting, immunodiffusion, immunoelectrophoresis, hemagglutination, and cell-binding assays.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4.

Analysis Note

The purity and identity are determined by immunoelectrophoresis, indirect ELISA, and SDS-PAGE.


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library