Skip to Content
MilliporeSigma

I4401

Interferon-αA/D human

≥95%, recombinant, expressed in E. coli, buffered aqueous solution, suitable for cell culture

Synonym(s):

IFN-αA/D

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human

Quality Segment

recombinant

expressed in E. coli

assay

≥95%

form

buffered aqueous solution

mol wt

19 kDa

packaging

pkg of 100K units

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

color

colorless

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... IFNA2(3440)

Application

Interferon-αA/D human has been used:
  • for identification of coronavirus replicase inhibitors
  • in combination with ACH-806 [1-(4-pentyloxy-3-trifluoromethylphenyl)-3-(pyridine-3-carbonyl)thiourea] to study the effect on hepatitis C virus (HCV) replication
  • in combination with vitamin D to check reduction in HCV protein production in cell culture

Biochem/physiol Actions

Interferon-αA/D (Bg/II), also known as Universal Type I Interferon, is an α interferon hybrid that crosses the species barrier. The Bg/II denotes the restriction enzyme site from which this hybrid is constructed from recombinant human interferons αA and αD. Interferon-αA/D has been proven to be active on many mammalian cells. It can substitute for human, monkey, mouse, bovine, rat, cat, dog, rabbit, sheep, goat, horse, pig or hamster type I interferon (α, β, ω and τ). Type I Interferons are a closely related family of 165-172 amino acid proteins that are produced by leukocytes (α subtypes), fibroblasts (β subtypes), lymphocytes (ω subtypes), and ruminant embryos (τ subtypes). This cytokine has anti-viral, anti-proliferative, immunoregulatory, and proinflammatory activities.
This cytokine has anti-viral, anti-proliferative, immunoregulatory, and proinflammatory activities.

Physical form

Solution in phosphate buffered saline containing 0.1% bovine serum albumin

Analysis Note

The biological activity is determined in the cytopathic effect inhibition assay using MDBK cells with vesicular stomatitis virus (VSV). A cytopathic effect of 50% is produced with 1 unit/ml interferon. The units are determined with respect to NIH international standard reference for human interferonαA.


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library