Skip to Content
MilliporeSigma

I4276

Interferon-αA human

≥95%, suitable for cell culture, buffered aqueous solution, recombinant, expressed in E. coli

Synonym(s):

IFN-αA, Interferon-α2A

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.32
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human

Quality Segment

recombinant

expressed in E. coli

assay

≥95%

form

buffered aqueous solution

mol wt

19 kDa

packaging

pkg of 20 μg

concentration

≥0.2 mg/mL

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

color

colorless

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IFNA2(3440)

Application

Interferon-αA human has been used to:
  • study the effect of Chikungunya virus on interferon-mediated JAK (Janus kinase)-STAT (signal transducers and activators of transcription) signaling
  • study the effect of interferon-α and PRK2 (protein kinase C-related kinase 2) inhibitors on hepatitis C virus replication
  • compare the effect of Monascus pigment derivatives on hepatitis C virus replication

Biochem/physiol Actions

Cytokine with anti-viral, anti-proliferative, immunoregulatory, and proinflammatory activities.
Cytokine with anti-viral, anti-proliferative, immunoregulatory, and proinflammatory activities. Type I Interferons are a closely related family of 165-172 amino acid proteins that are produced by leukocytes (α subtypes), fibroblasts (β subtypes), lymphocytes (ω subtypes) and ruminant embryos (τ subtypes). Interferon induces a variety of effects on target cells including antiviral, antiproliferative and immunomodulatory activities.

Physical form

Supplied as a solution in 5 mM sodium acetate, pH 4.0.

Analysis Note

The biological activity is determined in the cytopathic effect inhibition assay using MDBK cells with vesicular stomatitis virus (VSV). A cytopathic effect of 50% is produced with 1 unit/ml interferon. The units are determined with respect to WHO International standard reference for human interferonαA.


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library