Skip to Content
MilliporeSigma

Skip To

I3639

Isoetharine mesylate salt

Synonym(s):

4-[1-Hydroxy-2-[(1-methylethyl)amino]butyl]-1,2-benzenediol mesylate salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C13H21NO3 · CH4O3S
CAS Number:
Molecular Weight:
335.42
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
EC Number:
230-695-8
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


originator

Sanofi Aventis

Quality Segment

storage temp.

2-8°C

SMILES string

CS(O)(=O)=O.CCC(NC(C)C)C(O)c1ccc(O)c(O)c1

InChI

1S/C13H21NO3.CH4O3S/c1-4-10(14-8(2)3)13(17)9-5-6-11(15)12(16)7-9;1-5(2,3)4/h5-8,10,13-17H,4H2,1-3H3;1H3,(H,2,3,4)

InChI key

SOYAGMVKMXZVNZ-UHFFFAOYSA-N

Gene Information

Application

Isoetharine mesylate salt has been used:
  • as an adrenergic drug to test its effect on embryo implantation in 4-day pregnant mice[1]
  • as a substrate for recombinant human sulfotransferase (SULT) in SULT assay to monitor conjugation reaction[2]
  • as a β-2 agonist in collision-induced dissociation (CID) mass spectrometry analysis

Biochem/physiol Actions

Isoetharine is an agonist for β-adrenergic receptors and is a bronchodilator. It is a catecholic drug metabolized by sulfotransferases and is present in human urine and plasma as a sulfated metabolite.[2] Catechol O-methyltransferase (COMT) also mediates the methylation of isoetharine in the metabolic conjugation pathway.[2]

Features and Benefits

This compound was developed by Sanofi Aventis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
T2528B2134B2185
Quality Level

200

Quality Level

100

Quality Level

-

Quality Level

100

storage temp.

2-8°C

storage temp.

-

storage temp.

2-8°C

storage temp.

-

originator

Sanofi Aventis

originator

Novartis

originator

-

originator

Teva

Gene Information

human ... ADRB1(153), ADRB2(154), ADRB3(155)

Gene Information

human ... ADRB1(153), ADRB2(154), ADRB3(155)

Gene Information

human ... DRD2(1813), DRD3(1814), PRL(5617)

Gene Information

human ... ADRB1(153)


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters