Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I2765

Sigma-Aldrich

Ibotenic acid

~95% (TLC), solid, neurotoxin

Synonym(s):

α-Amino-3-hydroxy-5-isoxazoleacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6N2O4
CAS Number:
Molecular Weight:
158.11
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.32

Product Name

Ibotenic acid, ~95%, solid

Assay

~95%

Quality Level

form

solid

color

white

solubility

H2O: 1 mg/mL, clear, colorless (with sonication)
0.1 M NaOH: 10.7 mg/mL
0.1 M HCl: 4.7 mg/mL

SMILES string

NC(C(O)=O)C1=CC(=O)NO1

InChI

1S/C5H6N2O4/c6-4(5(9)10)2-1-3(8)7-11-2/h1,4H,6H2,(H,7,8)(H,9,10)

InChI key

IRJCBFDCFXCWGO-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Ibotenic acid has been used as a neurontoxin in rats. It has also been used in inducing a primary permanent lesion monkey cortex M1 neurons.
Ibotenic acid has been used to perform surgery in adult male zebra finches (Taeniopygia guttata) to study reversing reinforcement-induced vocal changes.

Biochem/physiol Actions

Ibotenic acid is an analog of glutamate. It is involved in the excitation of cell bodies and destroys the thalamic reticular nucleus (NRT). It induces high calcium influx leading to neuronal injury. Ibotenic acid is used to induce neurodegeneration due to its excitotoxic functionality.
Non-selective agonist with preference for NMDA glutamate receptors; neurotoxin; neuroexcitatory amino acid originally isolated from Amanita species.

Caution

Hygroscopic

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Megan E Fox et al.
ACS chemical neuroscience, 7(12), 1681-1689 (2016-09-13)
Central norepinephrine signaling influences a wide range of behavioral and physiological processes, and the ventral bed nucleus of the stria terminalis (vBNST) receives some of the densest norepinephrine innervation in the brain. Previous work describes norepinephrine neurons as projecting primarily
Benjamin M Basile et al.
PLoS biology, 18(6), e3000677-e3000677 (2020-06-13)
A key feature of most social relationships is that we like seeing good things happen to others. Research has implicated the anterior cingulate cortex (ACC) in attaching value to social outcomes. For example, single neurons in macaque ACC selectively code
Anja T Zai et al.
Nature communications, 11(1), 5940-5940 (2020-11-25)
Sensory substitution is a promising therapeutic approach for replacing a missing or diseased sensory organ by translating inaccessible information into another sensory modality. However, many substitution systems are not well accepted by subjects. To explore the effect of sensory substitution
Jiachao Wang et al.
Behavioural brain research, 372, 112037-112037 (2019-06-17)
Our internal models of the world help us to process information rapidly: in general model-based learning is more rapid than model-free learning. However, the cognitive flexibility required to overcome cognitive predispositions can let us down: it is not fully developed
L T Lopes et al.
Acta physiologica (Oxford, England), 211(3), 528-537 (2014-03-13)
Although periaqueductal grey matter activation is known to elicit respiratory and cardiovascular responses, the role of this midbrain area in the compensatory responses to hypoxia is still unknown. To test the participation of the periaqueductal grey matter in cardiorespiratory and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service