Skip to Content
MilliporeSigma

I0764

Inoculating Loops, calibrated

loop diam. × L 10 μL × 70 mm

Synonym(s):

inoculating loop, inoculation loop, inoculation loops

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.22
UNSPSC Code:
41161502
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

19 gauge platinum wire with 15% Iridium

packaging

pkg of 1 ea

loop diam. × L

10 μL × 70 mm

General description

Inoculating loops, calibrated allows an inoculum to be taken from a liquid by using surface tension, without handle.


Still not finding the right product?

Explore all of our products under Inoculating Loops, calibrated




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library