Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.47
UNSPSC Code:
41116121
form
solution
Quality Segment
shelf life
Expiry date on the label
IVD
for in vitro diagnostic use
application(s)
hematology
histology
storage temp.
room temp
Application
Intended for use in the study of connective tissue, muscle and collagen fibers. Stains collagen blue and muscle red.
Other Notes
Chromotrope 2R, aniline blue and phosphotungstic acid.
Storage Class
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
PPE (personal protective equipment)
Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.