Key Documents
HS29025A
Microtube Rack
natural
About This Item
UNSPSC Code:
41122800
NACRES:
NB.12
material
natural
polypropylene
feature
to hold 80 tube(s)
packaging
pack of 5 ea
manufacturer/tradename
Heathrow Scientific HS29025A
L × W × H
225 mm × 67 mm × 28 mm
Related Categories
General description
The 80-Well Microtube Rack is a popular fraction-collector-plate style rack.
Features and Benefits
  • Easily fill and transfer 80 tubes in compact arrangement (5x16 array)
  • Frosted labeling area on bottom of rack
  • Embossed Index Numbers for quick sample ID and retrieval
  • Made of polypropylene
  • Autoclavable
Choose from one of the most recent versions:
Certificates of Analysis (COA)