Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.22
UNSPSC Code:
41123300
Manufacturer/tradename:
Heathrow Scientific HS2860T
Feature:
Holds 36 x 15 mL tubes
Material:
cardboard
Packaging:
pk of 10
material
cardboard
feature
Holds 36 x 15 mL tubes
packaging
pk of 10
manufacturer/tradename
Heathrow Scientific HS2860T
well diam.
23 mm
Features and Benefits
- low cost way to organize your tubes
- ecomonical cardboard racks are disposable
- write-on surface for descriptions or self numbering system
- partitions supplied flat - packed for space-saving storage
- partitions available in two sizes
Legal Information
TrueNorth is a registered trademark of Heathrow Scientific LLC
Still not finding the right product?
Explore all of our products under TrueNorth® Cardboard Storage Box Partition
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.