MilliporeSigma
HS234631
Pipette Workstation
clear (workstation)
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.01
UNSPSC Code:
41121800
Material:
acrylic , clear (workstation)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
acrylic , clear (workstation)
feature
holds 4 x pipettes and pipette tips
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific HS234631
L × W × H
244 mm × 165 mm × 246 mm
General description
Organize your pipettes and tips in the Pipette Workstation and keep pipettes and disposable tips together in one convenient stand.
Features and Benefits
  • Acrylic workstation is opaque on sides with clear front and bin lids for easy content identification
  • Holds up to four pipettes on top shelf
  • Dual bins hold multiple size tips
  • Self-closing lid provides dust-free storage
  • Oversized lip allows for easy access when wearing gloves
Still not finding the right product?
Explore all of our products under Pipette Workstation
Choose from one of the most recent versions:
Certificates of Analysis (COA)