Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HS2201B

Droplet phlebotomy tray inserts for 16 mm test tube rack

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122800
NACRES:
NB.35

feature

Holds 16 mm Tube Rack

packaging

pack of 4 inserts

manufacturer/tradename

Heathrow Scientific HS2201B

General description

Utility Tray Insert B for 16mm test tube rack with eight compartments.

Legal Information

Droplet is a trademark of Heathrow Scientific LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service