MilliporeSigma
HS15981B
Slide holder
for 78 slides, white polyethylene
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.35
UNSPSC Code:
41123302
Manufacturer/tradename:
Heathrow Scientific HS15981B
Feature:
Holds 78 x slides 76x26 mm
Material:
white polyethylene
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
white polyethylene
feature
Holds 78 x slides 76x26 mm
manufacturer/tradename
Heathrow Scientific HS15981B
size
299x178x21 mm
General description
The Slide Holder holds slides during evaluation, cleaning, and drying.
  • Made from chemical-resistant polyethylene
  • Solid, strong construction with rubber feet for bench top stability
Still not finding the right product?
Explore all of our products under Slide holder
Choose from one of the most recent versions:
Certificates of Analysis (COA)