Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HS1423AA

Weigh Boat

Small, square black polystyrene, Anti-static, pk of 500 ea

Synonym(s):

weigh boat, weigh dish, weighing dish, weighing pan

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121812
NACRES:
NB.35

material

square black polystyrene

size

small

feature

Anti-static

packaging

pk of 500 ea

manufacturer/tradename

Heathrow Scientific HS1423AA

L × W × H

(46 mm) × 46 mm × 8 mm

color

black

Looking for similar products? Visit Product Comparison Guide

General description

Flexible polystyrene boats feature smooth surfaces to provide accurate pour-outs with minimal sample loss.

Features and Benefits

  • biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols, and bases.
  • temperature range -10 to 70C
  • disposable
  • shallow with a wide, flat bottom to resist tipping
  • rounded corners simplify transfer


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service