Key Documents
HS120668
Box Top Dispenser for Parafilm
purple (dispenser)
About This Item
UNSPSC Code:
41122800
NACRES:
NB.51
material
acrylonitrile-butadiene-styrene (ABS)
purple (dispenser)
feature
to hold 2 x 50 mm or 1 x 100 mm Parafilm M Sealing Film roll
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific 120668
L × W × H
123 mm × 172 mm × 99 mm
General description
The Box Top Dispenser is an easy application with no mess.
Features and Benefits
  • Accommodates two 50 mm (2-inch), or one 100 mm (4-inch) Parafilm® M rolls
  • Easy application, simply place the dispenser on top of the Parafilm® M box and pull the sealing film through, there is no need to remove rolls from the packaging
  • Built in safety blade slides and cuts with no danger to fingertips and eliminates the need for scissors
  • Supplied with two safety blades, additional replacement blades are available for purchase
  • Made from ABS plastic making the unit easy to clean
Legal Information
Parafilm is a registered trademark of Bemis Company, Inc.