MilliporeSigma
HS120640
Reagent Reservoir
capacity 25 mL, polystyrene, sterile; electron beam irradiated, pack of 200 (packed as 5 ea)
Synonym(s):
Chemical Reservoir
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.01
UNSPSC Code:
41121522
Capacity:
25 mL
Material:
V-bottom bottom, polystyrene
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
V-bottom bottom, polystyrene
sterility
sterile; electron beam irradiated
packaging
pack of 200 (packed as 5 ea)
manufacturer/tradename
Heathrow Scientific 120640
capacity
25 mL
General description
Reagent Reservoirs offer a fresh and modern design with enhanced features to streamline pipetting procedures. Graduation markingson the top edge and inner walls allows measurements of liquid to be easily identified and reduces waste of expensive reagents. Pour-off spouts design on all four corners enables easy spill-free pouring and reduces chance of dripping.
Features and Benefits
  • Trough design with a “v” shaped bottom helps ensure that all the reagent is accessible and can be easily aspirated
  • Thick wall construction and wide base adds stability and durability making the reservoir easy to handle without product deformation which results in less spillage
  • Reservoirs can be conveniently stacked with a unique nesting design that enables easy separation
  • Designed to stay flat on the benchtop and not warp
  • Sealed in easy-to-open bags. No scissors needed to open
  • Disposable
  • Design Patent Pending
Still not finding the right product?
Explore all of our products under Reagent Reservoir
Choose from one of the most recent versions:
Certificates of Analysis (COA)