Key Documents
HS120585
Magnetic Induction Stirrer
AC/DC input 100 V / 240 V, universal plug set, gray-blue
About This Item
UNSPSC Code:
41103800
NACRES:
NB.72
material
gray-blue
silicone plate
packaging
pkg of 1 ea
manufacturer/tradename
Heathrow Scientific 120585
parameter
2,000 rpm speed (max.)
plate diam.
160 mm
General description
Strong, motor-less and maintenance free
Superior Functional Design
  • Advanced magnetic coil technology drives the spinning of the stir bar maintaining continuous, constant speed for reproducible results
  • Magnetic coupling is optimized by using alternating current (AC) as its driving force strong enough to stir viscous solutions
  • Motor-less operation generates minimal heat ideal for stirring temperature sensitive samples and environments for long periods
  • With no moving parts, produces less vibration and quieter operation over traditional magnetic stirrers
Intuitive Operation
  • Multiple stirring modes are easy to program
  • Bright digital display precisely controls time and speed
  • Quickly meets stirring speeds with rapid acceleration and braking (less than 10 seconds)
Packaging
INCLUDES:
Magnetic Induction Stirrer
2 Stir Bars 35 mm x 12 mm, Cylindrical Shape
1 Silicone Anti-skid Plate Cover
1 Low Voltage, Double Insulated Power Adapter (110-240 VAC, 50/60 Hz) with 4 Interchangeable Plugs