MilliporeSigma
HS120481
Universal carousel pipette stand
purple to clear, 8 of each color for top + 8 clear bottoms
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.51
UNSPSC Code:
41122801
eCl@ss:
32032209
Manufacturer/tradename:
Heathrow Scientific HS120481
Feature:
Holds 16 x pipettors
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
feature
Holds 16 x pipettors
manufacturer/tradename
Heathrow Scientific HS120481
color
purple to clear, 8 of each color for top + 8 clear bottoms
General description
Pipettes never looked so good, on your bench top
Hold and store pipettes in style with the new universal carousel pipette stand that holds up to 8 single channel, 4 multi channel, or a combination of both pipettes. A sleek and modern design enables this stand to securely hold virtually every known brand of pipette in the marketplace making it truly ‘universal′ while adding a pop of color to a bench top. Inserts on the upper holders can be added or removed to accommodate the fit of different of pipette widths. One handed removal or replacement of pipettes is made easy with a rotating head and stable weighted base. Moving the stand while keeping pipettes in place is simple with a quick grip handle allowing a user to grab and go. The pipette stands come with additional upper leaves to enable multiple color coding configuration options as well as a change in color to your work space, for a bit more fun.
  • Ideal for labs that do not wish to be tied into a specific brand of pipette, or already have multiple brands in use
  • Weighted base allows for easy removal or replacement of pipettes, with one hand
  • Pipettes are held in a vertical position to help prevent contamination
  • Conveniently rotate stand head to select or store pipettes
  • Quick grip handle to move pipette stand (loaded or unloaded)
  • Assorted color options allow for color coding pipette location: Blue/Green; Purple/Clear; Assorted/Clear
  • The unit is not autoclavable but can be cleaned with soap/water or common laboratory disinfectants
  • Made of ABS (Acrylonitrile Butadiene Styrene)
  • US Patent Pending
Still not finding the right product?
Explore all of our products under Universal carousel pipette stand
Choose from one of the most recent versions:
Certificates of Analysis (COA)