MilliporeSigma
HS120375
Droplet Sample Storage Tray
polypropylene (corrugated), holds 24 sample containers, pack of 10 trays
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.22
UNSPSC Code:
41122800
Material:
polypropylene (corrugated)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
polypropylene (corrugated)
packaging
pack of 10 trays
L × W × H
16 × 10.24 × 3.5 in.
size
24 wells
color
white
General description
Corrugated storage trays can be used with the included dividers or sample containers or remove them for general lab product storage and transport.
Features and Benefits
  • moisture resistant, significantly reducing chance of mold
  • lightweight and sturdy to hold up to 24 full sample containers
  • stackable with other trays
  • removable dividers allow outer shell to be used for storage or transport of laboratory products
  • pack includes 10 trays and 10 dividers
  • disposable
  • supplied flat-packed to reduce storage space
Legal Information
Droplet is a trademark of Heathrow Scientific LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)