Key Documents
HS120365
Lab Alert® 4 way timer with whiteboard and pen
About This Item
UNSPSC Code:
41122411
NACRES:
NB.51
manufacturer/tradename
Heathrow Scientific HS120365
General description
  • Clock feature count up or down to 24 hours 59 minutes
  • 4 timer channels
  • Stands up or attaches to a magnetic surface (magnets on back of unit)
  • Whiteboard and marking pen included
  • Sounds when time expires
  • Batteries included (2 AAA)
Legal Information
Lab Alert is a registered trademark of Heathrow Scientific