Documents
HS120212
Vortex Mixer
AC/DC input 230 V / 240 V, Schuko plug, blue
About This Item
UNSPSC Code:
41103814
NACRES:
NB.73
material
blue
packaging
pkg of 1 ea
manufacturer/tradename
Heathrow Scientific 120212
General description
Multi-purpose, maximum convenience
One mixing head does it all with this sleek, modern and compact design vortex mixer!
Mixing Head Holding Capacity:
  • Center pad area for hand held mixing
  • Assay plate x 1 (to be used with retention cords)
  • 50 mL conical tubes x 2 (horizontally mounted)
  • 15 mL conical tubes x 2 (horizontally mounted)
  • 5 mL tubes x 2 (horizontally mounted in the same retention space as 15 mL)
  • 1.5/2 mL microfuge tubes x 4 (vertically mounted)
  • 0.5 mL microfuge tubes x 6 (vertically mounted)
  • 0.2 mL microfuge tubes (strip tubes) x 24 (vertical mounted)
Control switch with three operation selections (intermittent, off, continuous).
Infinite position rotary speed control from Off up to approx 3000 rpm
Made of durable, easy to clean ABS material and stable elastomer feet stabilize the unit and dampen vibration.
Packaging
INCLUDES:
1 Mini Vortexer
2 Silicone Caps