Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

HPA069577

Sigma-Aldrich

Anti-DUSP1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody

Synonym(s):

CL100, HVH1, MKP-1, PTPN10

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL

immunogen sequence

QVLAPHCSAEAGSPAMAVLDRGTSTTTVFNFPVSIPVHSTNSALSYLQS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DUSP1(1843)

General description

Dual specificity phosphatase 1 (DUSP1) gene encodes for a protein phosphatase enzyme with dual specificity. It is majorly expressed in epithelial tumors, such as prostate, colon, and bladder. DUSP1 protein consists of two domains comprising an amino-terminal noncatalytic domain and a C-terminal catalytic domain, which has an adenosine triphosphatase (ATPase) active site consensus sequence. It belongs to the family of dual-specificity phosphatases. The DUSP1 gene is mapped to human chromosome 5q35.1.

Immunogen

dual specificity phosphatase 1

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Dual specificity phosphatase 1 (DUSP1) protein functions as an activator in the mitogen-activated protein kinase (MAPK) signaling pathway, which dephosphorylate MAP kinase/extracellular-signal-regulated kinase (ERK-2) on tyrosine-185 and threonine-183. This is implicated in signal transduction, protein modification, and oxidative stress in normal cells. DUSP1 regulates invasion, angiogenesis, and metastasis of non-small cell lung cancer. Increased levels of DUSP1 promoter hypermethylation may correlate to tumors and cancer.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST90274

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Goot Heah Khor et al.
International journal of medical sciences, 10(12), 1727-1739 (2013-10-25)
Hypermethylation in promoter regions of genes might lead to altered gene functions and result in malignant cellular transformation. Thus, biomarker identification for hypermethylated genes would be very useful for early diagnosis, prognosis, and therapeutic treatment of oral squamous cell carcinoma
Jiliang Shen et al.
Cancer medicine, 5(8), 2061-2068 (2016-05-27)
Dual-specificity phosphatase-1 (DUSP1/MKP1), as a member of the threonine-tyrosine dual-specificity phosphatase family, was first found in cultured murine cells. The molecular mechanisms of DUSP1-mediated extracellular signal-regulated protein kinases (ERKs) dephosphorylation have been subsequently identified by studies using gene knockout mice

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service